siRNA Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 12 | 1422 | ||
GenBank mRNA | AK004748.1 | - | 12 | 1514 | ||
AK011110.1 | - | 5 | 506 | |||
AK131635.1 | - | 12 | 1426 | |||
AK134327.1 | - | 12 | 1514 | |||
AK147074.1 | - | 12 | 1423 | |||
AK167309.1 | - | 12 | 1518 | |||
AK170882.1 | - | 12 | 1502 | |||
AK209366.1 | - | 2 | 93 | |||
BC002255.1 | - | 9 | 952 | |||
BC033271.1 | - | 12 | 1460 | 4016197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
