siRNA Details
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024218.4 | NP_077180.1 | 3, 4 | 352 | ||
GenBank mRNA | AK018731.1 | - | 3, 4 | 235 | ||
AK077617.1 | - | 3, 4 | 226 | |||
AK136631.1 | - | 3, 4 | 235 | |||
BC002110.1 | - | 3, 4 | 227 | |||
BC053377.1 | - | 3, 4 | 223 | 4216355 | ||
BC058114.1 | - | 3, 4 | 222 | 6817726 | ||
BC092008.1 | - | 3, 4 | 218 | 30612436 | ||
BY031373.1 | - | 3, 4 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
