siRNA Details
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024218.4 | NP_077180.1 | 6 | 571 | ||
GenBank mRNA | AK018731.1 | - | 6 | 454 | ||
AK077617.1 | - | 6 | 445 | |||
AK136631.1 | - | 6 | 454 | |||
BC002110.1 | - | 6 | 446 | |||
BC053377.1 | - | 6 | 442 | 4216355 | ||
BC058114.1 | - | 6 | 441 | 6817726 | ||
BC092008.1 | - | 6 | 437 | 30612436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
