siRNA Details
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025826.4 | NP_080102.1 | 4 | 584 | ||
GenBank mRNA | AK004889.1 | - | 4 | 485 | ||
AK076535.1 | - | 4 | 485 | |||
AK131628.1 | - | 4 | 531 | |||
BC011235.1 | - | 4 | 466 | |||
BC035231.1 | - | 4 | 450 | |||
BC044826.1 | - | 4 | 517 | |||
BC054428.1 | - | 4 | 526 | 5039627 |

More Information
Other Species Targeted

Gene Ontology Categories:
