siRNA Details
Gene Symbol: Kdelr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025841.4 | NP_080117.1 | 4 | 730 | ||
GenBank mRNA | AJ278133.1 | - | 4 | 723 | ||
AK003515.1 | - | 4 | 725 | |||
AK007403.1 | - | 4 | 714 | |||
AK150402.1 | - | 4 | 725 | |||
BC007146.1 | - | 4 | 712 | 3496891 | ||
CT010232.1 | - | 4 | 588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
