siRNA Details
Gene Symbol: Myeov2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163425.1 | NP_001156897.1 | 3 | 392 | ||
GenBank mRNA | AA764377.1 | - | 209 | |||
AK003315.1 | - | 3 | 373 | |||
AK012198.1 | - | 3 | 365 | |||
AK152318.1 | - | 3 | 359 | |||
BC049759.1 | - | 3 | 392 | |||
BY631936.1 | - | 3 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
