siRNA Details
Gene Symbol: Ndufb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025843.3 | NP_080119.1 | 3 | 409 | ||
GenBank mRNA | AK002678.1 | - | 3 | 407 | ||
AK003300.1 | - | 3 | 410 | |||
BC034787.1 | - | 3 | 388 | 4913656 | ||
BC056984.1 | - | 2 | 4250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
