siRNA Details
Gene Symbol: Rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025886.3 | NP_080162.3 | 3 | 698 | ||
GenBank mRNA | AK002243.1 | - | 3 | 638 | ||
AK010305.1 | - | 3 | 724 | |||
BC011131.1 | - | 2 | 625 | 4206694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
