siRNA Details
Gene Symbol: Khdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025890.3 | NP_080166.1 | 1, 2 | 272 | ||
GenBank mRNA | AB211060.1 | - | 1, 2 | 209 | ||
AK010377.1 | - | 1, 2 | 209 | |||
AK145595.1 | - | 1, 2 | 272 | |||
AK166731.1 | - | 1, 2 | 222 | |||
BC137808.1 | - | 1, 2 | 180 | 8860827 | ||
BC137809.1 | - | 1, 2 | 180 | 8860828 | ||
DQ917421.1 | - | 1, 2 | 160 | |||
DQ917422.1 | - | 1, 2 | 160 |
