siRNA Details
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144948.5 | NP_659197.2 | 5 | 883 | ||
GenBank mRNA | AF458961.1 | - | 5 | 847 | ||
AK005200.1 | - | 5 | 859 | |||
AK006755.1 | - | 5 | 848 | |||
AK008645.1 | - | 5 | 843 | |||
AK011223.1 | - | 5 | 865 | |||
AK145381.1 | - | 4 | 1706 | |||
AK151056.1 | - | 5 | 862 | |||
AK151827.1 | - | 5 | 882 | |||
AK153504.1 | - | 6 | 952 | |||
AK188365.1 | - | 1 | 310 | |||
AK190328.1 | - | 1 | 310 | |||
AK192901.1 | - | 1 | 313 | |||
BC054774.1 | - | 5 | 850 | 6807637 | ||
BC116931.1 | - | 5 | 832 | 40126250 | ||
BC119038.1 | - | 5 | 832 | 8733787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
