siRNA Details
Gene Symbol: Rbm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144948.5 | NP_659197.2 | 5 | 819 | ||
GenBank mRNA | AF458961.1 | - | 5 | 783 | ||
AK005200.1 | - | 5 | 795 | |||
AK006755.1 | - | 5 | 784 | |||
AK008645.1 | - | 5 | 779 | |||
AK011223.1 | - | 5 | 801 | |||
AK015662.1 | - | 5 | 821 | |||
AK145381.1 | - | 4 | 1642 | |||
AK151056.1 | - | 5 | 798 | |||
AK151827.1 | - | 5 | 818 | |||
AK153504.1 | - | 6 | 888 | |||
AK188365.1 | - | 1 | 246 | |||
AK190328.1 | - | 1 | 246 | |||
BC054774.1 | - | 5 | 786 | 6807637 | ||
BC116931.1 | - | 5 | 768 | 40126250 | ||
BC119038.1 | - | 5 | 768 | 8733787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
