siRNA Details
Gene Symbol: Gm11468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB870378.1 | - | 2, 3 | 305 | ||
BB872766.1 | - | 2, 3 | 306 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB870378.1 | - | 2, 3 | 305 | ||
BB872766.1 | - | 2, 3 | 306 |