siRNA Details
Gene Symbol: Ctnnbip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141930.1 | NP_001135402.1 | 4 | 553 | ||
NM_023465.4 | NP_075954.1 | 5 | 659 | |||
GenBank mRNA | AB021261.1 | - | 5 | 576 | ||
AK003595.1 | - | 4 | 555 | |||
AK009066.1 | - | 4 | 480 | |||
AK036181.1 | - | 5 | 661 | |||
AK048258.1 | - | 5 | 657 | |||
BC018436.1 | - | 5 | 619 | |||
BC038253.1 | - | 5 | 635 | 4974951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
