siRNA Details
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175114.3 | NP_780323.2 | 4 | 574 | ||
GenBank mRNA | AK029123.1 | - | 5 | 972 | ||
AK041436.1 | - | 5 | 942 | |||
AK052264.1 | - | 5 | 961 | |||
AK122426.1 | - | 5 | 664 | |||
BC046285.1 | - | 4 | 564 | |||
BC058971.1 | - | 4 | 516 | 6832507 | ||
BC116262.1 | - | 4 | 541 | |||
BC116263.1 | - | 4 | 541 | |||
BC129961.1 | - | 4 | 541 | |||
BC129962.1 | - | 4 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
