siRNA Details
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164290.1 | NP_001157762.1 | 5 | 444 | ||
NM_001164291.1 | NP_001157763.1 | 4 | 336 | |||
NM_001164292.1 | NP_001157764.1 | 4 | 333 | |||
NM_001164293.1 | NP_001157765.1 | 4 | 336 | |||
NM_001164294.1 | NP_001157766.1 | 4 | 336 | |||
NM_001164295.1 | NP_001157767.1 | 4 | 333 | |||
NM_024192.2 | NP_077154.2 | 5 | 435 | |||
GenBank mRNA | AB232359.1 | - | 3 | 234 | ||
AK003457.1 | - | 4 | 291 | |||
AK008842.1 | - | 4 | 299 | |||
AK013884.1 | - | 4 | 294 | |||
AK186231.1 | - | 4 | 259 | |||
AK207375.1 | - | 5 | 361 | |||
BC003442.1 | - | 5 | 399 | 2650719 | ||
BY279028.1 | - | 4 | 335 | |||
BY312150.1 | - | 4 | 297 | |||
CK625575.1 | - | 4 | 262 | |||
CX223880.1 | - | 5 | 400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
