siRNA Details
Gene Symbol: Rnf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025999.3 | NP_080275.2 | 6 | 683 | ||
GenBank mRNA | AF353167.1 | - | 6 | 713 | ||
AK006791.1 | - | 6 | 723 | |||
AK011839.1 | - | 6 | 672 | |||
AK015505.1 | - | 6 | 684 | |||
AK154417.1 | - | 6 | 707 | |||
BC018553.1 | - | 6 | 677 | 3582546 |

More Information
Other Species Targeted

Gene Ontology Categories:
