siRNA Details
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026001.2 | NP_080277.1 | 8 | 645 | ||
GenBank mRNA | AK011914.1 | - | 8 | 684 | ||
AK082752.1 | - | 8 | 646 | |||
BC047997.1 | - | 8 | 646 | 6510494 |

More Information
Other Species Targeted

Gene Ontology Categories:
