siRNA Details
Gene Symbol: Ndufa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023312.2 | NP_075801.1 | 5 | 1225 | ||
GenBank mRNA | AF286698.1 | - | 4 | 460 | ||
AK002999.1 | - | 4 | 464 | |||
AK003012.1 | - | 4 | 478 | |||
AK010517.1 | - | 4 | 464 | |||
AK012422.1 | - | 4 | 464 | |||
BC034899.1 | - | 5 | 1225 | 1447015 | ||
BC037149.1 | - | 4 | 468 | 5149812 | ||
BC055435.1 | - | 4 | 474 | 4219489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
