siRNA Details
Gene Symbol: Zkscan14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023322.2 | NP_075811.2 | 1, 2 | 607 | ||
GenBank mRNA | AK013258.1 | - | 1, 2 | 595 | ||
AK153972.1 | - | 1, 2 | 590 | |||
BC141244.1 | - | 1, 2 | 533 | 9055856 | ||
U62906.1 | - | 1, 2 | 577 |
