siRNA Details
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026066.2 | NP_080342.1 | 2 | 383 | ||
GenBank mRNA | AK005375.1 | - | 2 | 338 | ||
AK013666.1 | - | 2 | 383 | |||
AY241867.1 | - | 2 | 373 | |||
BC049665.1 | - | 2 | 370 | 6533928 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
