siRNA Details
Gene Symbol: Pagr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030240.1 | NP_084516.1 | 3 | 1373 | ||
GenBank mRNA | AK013840.1 | - | 3 | 1347 | ||
AK082775.1 | - | 3 | 1082 | |||
AK179443.1 | - | 1 | 192 | |||
AK219667.1 | - | 1 | 192 | |||
BC003932.1 | - | 3 | 1373 | 3485982 | ||
BC054744.1 | - | 4510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
