siRNA Details
Gene Symbol: 3110040N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026077.3 | NP_080353.1 | 2, 3 | 229 | ||
GenBank mRNA | AK014163.1 | - | 2, 3 | 209 | ||
AK018003.1 | - | 2, 3 | 255 | |||
AK019261.1 | - | 2, 3 | 229 | |||
AK170168.1 | - | 2, 3 | 246 | |||
BC027500.1 | - | 2, 3 | 217 | 1448876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
