siRNA Details
Gene Symbol: Ccdc137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152807.3 | NP_690020.1 | 2 | 198 | ||
GenBank mRNA | AK156682.1 | - | 2 | 207 | ||
BC026662.1 | - | 2 | 200 | 4989084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
