siRNA Details
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025584.2 | NP_079860.2 | 502 | |||
GenBank mRNA | AB122023.1 | - | 502 | |||
AK004342.1 | - | 504 | ||||
AK010597.1 | - | 486 | ||||
AK044051.1 | - | 492 | ||||
AY262355.1 | - | 475 | ||||
BC019482.1 | - | 500 | 4218841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
