siRNA Details
Gene Symbol: Lyzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026092.3 | NP_080368.1 | 3 | 374 | ||
GenBank mRNA | AK006241.1 | - | 3 | 375 | ||
AK006630.1 | - | 3 | 375 | |||
AY572447.1 | - | 3 | 333 | |||
BC048450.1 | - | 3 | 345 | 6774504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
