siRNA Details
Gene Symbol: Tmed11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026109.2 | NP_080385.2 | 5 | 569 | ||
GenBank mRNA | AK007381.1 | - | 5 | 578 | ||
AK019032.1 | - | 5 | 558 | |||
BC132664.1 | - | 5 | 541 | 40130941 | ||
BC132666.1 | - | 5 | 541 | 40130943 | ||
BY707479.1 | - | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
