siRNA Details
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114087.2 | NP_001107559.1 | 8 | 988 | ||
NM_026131.4 | NP_080407.3 | 8 | 886 | |||
GenBank mRNA | AK003019.1 | - | 9 | 963 | ||
AK003338.1 | - | 8 | 877 | |||
AK010141.1 | - | 8 | 875 | |||
AK049902.1 | - | 5 | 1892 | |||
AK077998.1 | - | 8 | 980 | |||
AK128987.1 | - | 7 | 1182 | |||
BC049565.1 | - | 8 | 874 | 6588160 | ||
BC052698.1 | - | 8 | 959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
