siRNA Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 9 | 2499 | ||
NM_207232.2 | NP_997115.1 | 8 | 2316 | |||
GenBank mRNA | AB185283.1 | - | 1 | 100 | ||
AK030415.1 | - | 8 | 2309 | |||
AK038922.1 | - | 8 | 2229 | |||
AK132781.1 | - | 9 | 2408 | |||
BC066081.1 | - | 8 | 2217 | 30355705 | ||
BC068267.1 | - | 1 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
