siRNA Details
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026160.4 | NP_080436.1 | 3 | 228 | ||
GenBank mRNA | AF255953.1 | - | 3 | 246 | ||
AK002795.1 | - | 3 | 231 | |||
AK003106.1 | - | 3 | 229 | |||
AK003205.1 | - | 3 | 229 | |||
AK003558.1 | - | 3 | 231 | |||
AK012604.1 | - | 3 | 232 | |||
AK132329.1 | - | 236 | ||||
AK151614.1 | - | 3 | 227 | |||
BC068180.1 | - | 3 | 231 | 30634082 | ||
BC086774.1 | - | 2 | 2655 | |||
BC110308.1 | - | 3 | 219 | 5319360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
