siRNA Details
Gene Symbol: C1qtnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026161.3 | NP_080437.2 | 2 | 944 | ||
GenBank mRNA | AK002948.1 | - | 2 | 871 | ||
AK004340.1 | - | 1 | 493 | |||
AK029777.1 | - | 1 | 119 | |||
BC027315.1 | - | 2 | 792 | |||
BC087832.1 | - | 1 | 4256 | |||
DQ002397.1 | - | 1 | 721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
