siRNA Details
Gene Symbol: Pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026176.3 | NP_080452.2 | 4 | 745 | ||
GenBank mRNA | AK004704.1 | - | 4 | 1458 | ||
AK028668.1 | - | 4 | 635 | |||
AK028814.1 | - | 4 | 635 | |||
AK133747.1 | - | 4 | 655 | |||
AK166983.1 | - | 4 | 635 | |||
BC006578.1 | - | 4 | 645 | 3586612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
