siRNA Details
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 14, 15 | 1194 | ||
GenBank mRNA | BQ442103.1 | - | 7, 8 | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
