siRNA Details
Gene Symbol: Ndufc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024220.2 | NP_077182.1 | 3 | 543 | ||
GenBank mRNA | AK002373.1 | - | 3 | 451 | ||
AK003380.2 | - | 3 | 445 | |||
AK008502.1 | - | 3 | 458 | |||
AK008889.1 | - | 3 | 451 | |||
AK019002.1 | - | 3 | 470 | |||
AK153547.1 | - | 3 | 473 | |||
AK166594.1 | - | 3 | 470 | |||
AK168342.1 | - | 3 | 495 | |||
BC002097.1 | - | 3 | 466 | 3497384 | ||
CK128102.1 | - | 3 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
