siRNA Details
Gene Symbol: Gcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026572.3 | NP_080848.1 | 2 | 188 | ||
GenBank mRNA | AK003788.2 | - | 2 | 185 | ||
AK019983.1 | - | 2 | 189 | |||
AK050373.1 | - | 2 | 201 | |||
BC014745.1 | - | 2 | 192 | 4239181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
