siRNA Details
Gene Symbol: Lrriq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290510.1 | NP_001277439.1 | 5 | 1908 | ||
NM_026668.2 | NP_080944.2 | 7 | 2071 | |||
GenBank mRNA | AK015024.1 | - | 5 | 1859 | ||
AK015586.1 | - | 1 | 83 | |||
AK019775.1 | - | 7 | 2072 | |||
BC145806.1 | - | 7 | 1772 | 40131109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
