siRNA Details
Gene Symbol: 1110007C09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026738.2 | NP_081014.1 | 6 | 658 | ||
GenBank mRNA | AK003526.1 | - | 6 | 659 | ||
AK154517.1 | - | 6 | 630 | |||
BC023485.1 | - | 7 | 790 | |||
BC038930.1 | - | 2 | 538 | |||
BC047205.1 | - | 6 | 698 | |||
BC147073.1 | - | 6 | 614 | 9056051 | ||
BC147074.1 | - | 6 | 614 | 9056042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
