siRNA Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037742.2 | NP_001032831.2 | 7 | 830 | ||
GenBank mRNA | BC023868.1 | - | 7 | 822 | ||
BC058590.1 | - | 6 | 1146 | |||
BC099495.1 | - | 7 | 572 | |||
BC132289.1 | - | 7 | 679 | 40130566 | ||
BC144765.1 | - | 7 | 645 | |||
BC144766.1 | - | 7 | 660 | |||
CF951173.1 | - | 8 | 606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
