siRNA Details
Gene Symbol: 1500012K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005235.1 | - | 1, 2 | 223 | ||
AK035406.1 | - | 1, 2 | 216 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1500012K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005235.1 | - | 1, 2 | 223 | ||
AK035406.1 | - | 1, 2 | 216 |