siRNA Details
Gene Symbol: Hddc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027168.2 | NP_081444.1 | 4 | 356 | ||
GenBank mRNA | AK009957.1 | - | 4 | 297 | ||
BC104360.1 | - | 4 | 356 | 40049707 | ||
BC104361.1 | - | 4 | 356 | 40049709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
