siRNA Details
Gene Symbol: Pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197992.1 | NP_932109.1 | 5 | 484 | ||
GenBank mRNA | BC028560.1 | - | 5 | 484 | 1380201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
