siRNA Details
Gene Symbol: Ufsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027356.2 | NP_081632.1 | 1 | 641 | ||
GenBank mRNA | AK012343.1 | - | 1 | 642 | ||
BC016577.1 | - | 1 | 650 | 3584484 | ||
BC087958.1 | - | 1 | 662 | 6751787 |

More Information
Other Species Targeted

Gene Ontology Categories:
