siRNA Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027470.3 | NP_081746.1 | 5 | 1355 | ||
GenBank mRNA | AK088512.1 | - | 5 | 1332 | ||
AK129298.2 | - | 5 | 1351 | |||
AY217016.1 | - | 3 | 1070 | |||
BC030389.1 | - | 2 | 513 | |||
BC048238.1 | - | 5 | 1317 | 6475114 |
