siRNA Details
Gene Symbol: 4921524L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027598.1 | NP_081874.1 | 10 | 1243 | ||
GenBank mRNA | AK014961.1 | - | 10 | 1244 | ||
AK161257.1 | - | 625 | ||||
BC049627.1 | - | 8 | 1088 | 6706381 |
