siRNA Details
Gene Symbol: Zfp935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136496.1 | NP_001129968.1 | 2 | 203 | ||
NM_178875.4 | NP_849206.2 | 2 | 203 | |||
GenBank mRNA | AK018441.1 | - | 2 | 194 | ||
AY189975.1 | - | 2 | 179 | |||
BC012524.1 | - | 2 | 179 | 4008347 | ||
BC037448.1 | - | 2 | 208 | |||
BC047396.1 | - | 2 | 180 | 3979342 | ||
BC087936.1 | - | 2 | 203 | |||
BC113193.1 | - | 2 | 199 | 40062446 | ||
BC115966.1 | - | 122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
