siRNA Details
Gene Symbol: 1700007B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164235.1 | NP_001157707.1 | 11 | 1535 | ||
NM_027944.1 | NP_082220.1 | 10 | 1417 | |||
GenBank mRNA | AK005693.1 | - | 10 | 1418 | ||
AK077102.1 | - | 12 | 1274 | |||
BC117767.1 | - | 11 | 1438 | 40090978 | ||
BC117768.1 | - | 10 | 1320 | 40090984 |
