siRNA Details
Gene Symbol: Ppapdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293703.1 | NP_001280632.1 | 2, 3 | 209 | ||
NM_028000.1 | NP_082276.1 | 2, 3 | 209 | |||
GenBank mRNA | AA184457.1 | - | 2, 3 | 209 | ||
AK009455.1 | - | 2, 3 | 176 | |||
AK144593.1 | - | 2, 3 | 194 | |||
BB857717.1 | - | 2, 3 | 210 | |||
BC099489.1 | - | 2, 3 | 160 |
