siRNA Details
Gene Symbol: Jsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028001.3 | NP_082277.2 | 5 | 390 | ||
GenBank mRNA | AK009016.1 | - | 5 | 384 | ||
AK009578.1 | - | 5 | 605 | |||
BC104136.1 | - | 4 | 304 | |||
BC104137.1 | - | 4 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
