siRNA Details
Gene Symbol: 2310047M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028005.3 | NP_082281.2 | 1 | 1382 | ||
GenBank mRNA | AK151629.1 | - | 1 | 1384 | ||
AK169875.1 | - | 1 | 1363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
