siRNA Details
Gene Symbol: Tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028339.1 | NP_082615.1 | 1 | 157 | ||
GenBank mRNA | AK013150.1 | - | 1 | 157 | ||
AK030429.1 | - | 1 | 175 | |||
AK050567.1 | - | 1 | 156 | |||
AK137059.1 | - | 1 | 154 | |||
AK150511.1 | - | 1 | 164 | |||
AK153027.1 | - | 1 | 160 | |||
AK170468.1 | - | 1 | 175 | |||
BC017603.1 | - | 1 | 157 | 4503129 | ||
BC021533.1 | - | 1 | 178 | 5354337 |

More Information
Other Species Targeted

Gene Ontology Categories:
