siRNA Details
Gene Symbol: Fam76b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176836.3 | NP_789806.1 | 4, 5 | 612 | ||
GenBank mRNA | AK052430.1 | - | 4, 5 | 570 | ||
AK145026.1 | - | 4, 5 | 624 | |||
BC043120.1 | - | 4, 5 | 612 | 6408908 | ||
BC050870.1 | - | 4, 5 | 533 | 6335538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
